Order Entry
Japan
ContactUsLinkComponent
Acetic acid glacial ≥99.7%, BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Corrosive
Acetic acid glacial ≥99.7%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Flammable
Corrosive
Acetic acid glacial ≥99.7%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • ACS - Assay (by GC, corrected for water):
    ≥99.7 %
  • ACS - Color (APHA):
    ≤10
  • Dilution Test:
    Passes Test
  • ACS - Residue after Evaporation:
    ≤0.001 %
  • ACS - Acetic Anhydride ((CH₃CO)₂O):
    ≤0.01 %
  • ACS - Chloride (Cl):
    ≤1 ppm
  • ACS - Sulfate (SO₄):
    ≤1 ppm
  • ACS - Substances Reducing Dichromate:
    Passes Test
  • ACS - Substances Reducing Permanganate:
    Passes Test
  • ACS - Titrable Base (meq/g):
    ≤0.0004
  • ACS - Suitability for Cholesterol Determination:
    Passes Test
  • ACS - Acetaldehyde:
    ≤0.005 %
  • ACS - Heavy Metals (by ICP-OES):
    ≤5 ppm
  • Trace Impurities - ACS - Iron (Fe):
    ≤0.200 ppm

Specification Test Results

Related Information

Frequently Bought Together