


1-Butanol ≥99.4%, PHOTREX® for spectrophotometry, J.T.Baker®
9189-01
: Avantor
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)
- ACS - Assay (CH₃CH₂CH₂CH₂OH) (by GC, corrected for water):≥99.4 %
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤0.0005 %
- ACS - Titrable Acid (µeq/g):≤0.8
- ACS - Butyl Ether (C₈H₁₈O)(by GC):≤0.2 %
- Carbonyl Compounds - Butyraldehyde:≤0.01 %
- Water (by KF, coulometric):≤0.05 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-300 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:≤0.04
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.50
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 215 nm:≤1.00
:
:74.12 g/mol
:13,01539
:117.6 °C (1013 hPa)
:–89 °C
:0.812 g/cm³ (20 °C)
:30 °C
:Ambient
:200-751-6
:1120
:3,III
:Management of Change (MOC) category = N
:01-2119484630-38
:GHS02
:GHS05
:GHS07
:71-36-3
:H₃C(CH₂)₂CH₂OH
:MFCD00002964