Potassium chlorate ≥99.0% (by KMnO₄ titration), granular, AR® ACS, Macron Fine Chemicals™
MACR6834-04
:
- Pack type:Glass bottle
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
- ACS - Assay (KClO₃) (by KMnO₄ titrn):≥99.0 %
- ACS - Bromate (BrO₃):≤0.015 %
- ACS - Calcium (Ca):≤0.002 %
- ACS - Chloride (Cl):≤0.001 %
- ACS - Heavy Metals (as Pb):≤5 ppm
- ACS - Insoluble Matter:≤0.005 %
- ACS - Iron (Fe):≤3 ppm
- ACS - Magnesium (Mg):≤0.002 %
- ACS - Sodium (Na):≤0.01 %
- ACS - Sulfate (SO₄):Passes Test
:
:122.55 g/mol
:2.30 g/cm³
:5.1,II
:Management of Change (MOC) category = N
:1485
:GHS03
:GHS07
:GHS09
:3811-04-9
:KClO₃
:MFCD00011361