
L-Tyrosine disodium salt dihydrate ≥99% (by HPLC), Biotech Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:122666-87-9
:MFCD08276422
Formula:C₉H₉NO₃Na₂·2H₂O
MDL Number:MFCD03840558
:261.19 g/mol
:Management of Change (MOC) category = R
:GHS07
Specification Test Results
- Appearance (White to beige powder with occasional brown solid):Passes Test
- Identification (IR):Passes Test
- Solution (5%) in water:Passes Test
- Specific Rotation @ 20 °C:°
- Assay (by HPLC):≥ 99 %
- Water (by KF):≤ 19.3 %
- Bacterial endotoxins, EU/g:≤ 50
- Arsenic (As), For Information Only:ppm
- Cadmium (Cd), For Information Only:ppm
- Mercury (Hg), For Information Only:ppm
- Cobalt (Co), For Information Only:ppm
- Nickel (Ni), For Information Only:ppm
- Vanadium (V), For Information Only:ppm
- Silver (Ag), For Information Only:ppm
- Iridium (Ir), For Information Only:ppm
- Chromium (Cr), For Information Only:ppm
- Copper (Cu), For Information Only:ppm
- Iron (Fe), For Information Only:ppm
- Lithium (Li), For Information Only:ppm
- Molybdenum (Mo), For Information Only:ppm
- Antimony (Sb), For Information Only:ppm
- Tin (Sn), For Information Only:ppm
- Aluminum (Al), For Information Only:ppm
- Manganese (Mn), For Information Only:ppm
- Residual Solvents: Class 3, each:≤ 5000 ppm
- Total Aerobic Microbial Count:≤ 100 CFU/g
- Total Yeasts and Molds:≤ 100 CFU/g
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_BAKR2098
SDS
spec_BAKR2098-01 spec_BAKR2098-07
Product specification
Frequently Bought Together