Order Entry
Japan
ContactUsLinkComponent
Guanidinium hydrochloride solution 6 M, GenAR®, Macron Fine Chemicals™
Irritant
Guanidinium hydrochloride solution 6 M, GenAR®, Macron Fine Chemicals™
Catalog # MACR0570-26
CAS Number:  
Irritant
Guanidinium hydrochloride solution 6 M, GenAR®, Macron Fine Chemicals™
Catalog # MACR0570-26
CAS Number:  

Specifications

  • Pk:
    1.135 L
  • Seq:
    5849

Specifications

Specification Test Results

  • Molarity (by titrimetry):
    5.8 - 6.2
  • Appearance (clear, colorless liquid):
    Passes Test
  • Heavy Metals (as Pb):
    ≤10 ppm
  • Identification (by FTIR):
    Passes Test
  • ACS - Iron (Fe):
    ≤3 ppm
  • pH (1:10 dilution):
    4.5 - 6.5
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:
    ≤0.1
  • Cyanide (CN):
    Passes Test
  • Solution Test:
    Passes Test

Specification Test Results

About this item

 :
 :95.53 g/mol
Melting Pt:100 °C
EINECS:233-151-8
 :GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026