

Acetic acid 80% solution, Biotech Reagent, J.T.Baker®
Catalog # 0321-09
Supplier: Avantor
CAS Number:
Specifications
- Pk:200 L
- Seq:5549
Specifications
Specification Test Results
- Appearance:Passes Test
- Assay (CH₃COOH):78.0 - 82.0 %
- Chloride (Cl):≤25 ppm
- ACS - Heavy Metals (as Pb):≤5 ppm
- Iron (Fe):≤5 ppm
- Nonvolatile Residue (USP), mg/20 mL:≤1.0
- Sulfate (SO₄):Passes Test
Specification Test Results
About this item
:
:60.05 g/mol
UN:2790
ADR:8,II
:GHS05
:GHS07
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152