Order Entry
Japan
ContactUsLinkComponent
Sodium chloride 99.0-100.5% (dry basis), granular FCC, USP, J.T.Baker®
Irritant
Sodium chloride 99.0-100.5% (dry basis), granular FCC, USP, J.T.Baker®
Catalog # 3628-X2
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Sodium chloride 99.0-100.5% (dry basis), granular FCC, USP, J.T.Baker®
Catalog # 3628-X2
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    94,662 kg
  • Seq:
    4544

Specifications

Specification Test Results

  • Kit Components Meet Requirements:
    Passes Test

Specification Test Results

About this item

 :
 :58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³
 :Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:231-598-3
 :GHS07
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477