Order Entry
Japan
ContactUsLinkComponent
di-Potassium hydrogen phosphate kit 98.0-100.5% (dried basis) USP, J.T.Baker®
di-Potassium hydrogen phosphate kit 98.0-100.5% (dried basis) USP, J.T.Baker®
Catalog # 3251-X1
Supplier:  Avantor
CAS Number:  
undefined
di-Potassium hydrogen phosphate kit 98.0-100.5% (dried basis) USP, J.T.Baker®
Catalog # 3251-X1
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

Specification Test Results

  • Kit Components Meet Requirements:
    Passes Test

Specification Test Results

About this item

 :
 :174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383