Order Entry
Japan
ContactUsLinkComponent
 
di-Potassium hydrogen phosphate 98.0-100.5% (dried basis) USP, J.T.Baker®
Catalog # 3251-B1
Supplier:  Avantor
CAS Number:  
di-Potassium hydrogen phosphate 98.0-100.5% (dried basis) USP, J.T.Baker®
Catalog # 3251-B1
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    62,869 kg
  • Seq:
    4472

Specifications

Specification Test Results

  • USP - Assay (K₂HPO₄) (dried basis):
    98.0 - 100.5 %
  • USP - Arsenic (As):
    ≤3 ppm
  • USP - Carbonate (CO₃):
    Passes Test
  • USP - Chloride (Cl):
    ≤0.03 %
  • USP - Fluoride (F):
    ≤0.001 %
  • USP - Identification:
    Passes Test
  • USP - Insoluble Substances:
    ≤0.2 %
  • USP - Iron (Fe):
    ≤0.003 %
  • USP - Loss on Drying:
    ≤1.0 %
  • USP - Mono- or Tri- Basic Salt:
    Passes Test
  • USP - pH (1 in 20):
    8.5 - 9.6
  • USP - Sodium (Na):
    Passes Test
  • USP - Sulfate (SO₄):
    ≤0.1 %

Specification Test Results

About this item

 :
 :174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383