Potassium dihydrogen phosphate kit 98.0-100.5% (dried basis), crystals NF, Multi-Compendial, J.T.Baker®
Catalog # 3248-X1
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specification Test Results
About this item
:
:136.09 g/mol
Merck Index:13,07744
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
:GHS07
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401