VWR®, Safety Caps
VWR® safety cap extracts solvents or mobile phases safely from the bottle or reservoirs without letting the hazardous vapour come out. It filters particulates in the air and prevents contaminants and dusts entering solvents.
- Advanced vapour valve keeps lab environment free from hazardous solvent vapour
- Efficient PTFE filter for preventing contaminants mediated noise
- Specially designed O-ring for air tight connection with glass bottle
- Freely rotating cap body prevents twisting tubes during solvent bottle exchange
Safety Caps are recommended to use with HPLC solvents bottles for safer lab environment and lesser uncertainty of analysis.
Available with different number of ports. E.g. 1, 2, 3, 4 and 5.
: One box contains set of four caps and their accessories.
A new box will have 4 sets of port blocks (with hole and without hole) in it.
: The valve is recommended to be replaced after six months of use.
VWR® safety cap extracts solvents or mobile phases safely from the bottle or reservoirs without letting the hazardous vapour come out. It filters particulates in the air and prevents contaminants and dusts entering solvents.
- Advanced vapour valve keeps lab environment free from hazardous solvent vapour
- Efficient PTFE filter for preventing contaminants mediated noise
- Specially designed O-ring for air tight connection with glass bottle
- Freely rotating cap body prevents twisting tubes during solvent bottle exchange
Safety Caps are recommended to use with HPLC solvents bottles for safer lab environment and lesser uncertainty of analysis.
Available with different number of ports. E.g. 1, 2, 3, 4 and 5.
: One box contains set of four caps and their accessories.
A new box will have 4 sets of port blocks (with hole and without hole) in it.
: The valve is recommended to be replaced after six months of use.
Product Family Options
- DescriptionThreadConnector
- Safety cap with filter valveGL 451 port and 1 filter1 * 4 items$89.80
- Safety cap with filter valveGL 452 port and 1 filter1 * 4 items$89.80
- Safety cap with filter valveGL 453 port and 1 filter1 * 4 items$89.80
- Safety cap with filter valveGL 454 port and 1 filter1 * 4 items$89.80
- Safety cap with filter valveGL 455 port and 1 filter1 * 4 items$89.80