Order Entry
Japan
ContactUsLinkComponent
VWR®, Residual Solvents Class 1 According to Ph. Eur. Versions 8.8 and 10
VWR®, Residual Solvents Class 1 According to Ph. Eur. Versions 8.8 and 10
Catalog # 88215.001
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Residual Solvents Class 1 According to Ph. Eur. Versions 8.8 and 10
Catalog # 88215.001
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    Residual Solvents Class 1 acc. to Ph. Eur. 8.8
Specifications
About this item
The level of volatile organic solvents in pharmaceutical raw materials or finished product is defined and monitored. Residual solvents are classified according to their risk assessment classes. A selection of residual solvent standards is now available from Avantor.
Residual Solvents Class 1 acc to Ph Eur Ver 8.8: 5 components; benzene 4 mg/ml; 1,2-dichloroethane 10 mg/ml ; tetrachloromethane 8 mg/ml; 1,1-dichloroethene 16 mg/ml; 1,1,1-trichloroethane 20 mg/ml in imethylsulphoxide
Residual Solvents Class 1 acc. to Ph Eur Ver 10: 5 components; benzene 2 mg/L; tetrachloromethane (carbon tetrachloride) 4 mg/L; 1,2-dichloroethane 5 mg/L; 1,1-dichloroethene 8 mg/L; 1,1,1-trichloroethane 1500 mg/L in dimethyl sulphoxide.
Certifications: All Solutions are Certified Reference Materials, produced and calibrated under a quality system that is:
- ISO 9001 certified.
- accredited according to ISO/IEC 17025 - Testing
- accredited according to ISO 17034 - Reference Material Producer