Order Entry
Japan
ContactUsLinkComponent
BROXO salt, granules 6 - 15 mm
BROXO salt, granules 6 - 15 mm
Catalog # 5254.9025
Supplier:  VWR Chemicals
CAS Number:  
undefined
BROXO salt, granules 6 - 15 mm
Catalog # 5254.9025
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bag
  • Pk:
    25 kg
  • Seq:
    4283
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Saline, NaCl, Sea salt, Halite
 :
 :58.44 g/mol
Boiling Pt:1413
Melting Pt:801
Density:2.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477
Merck Index:14,08599