Order Entry
Japan
ContactUsLinkComponent
Toluene-α-sulphonyl fluoride ≥99.0%, Proteomics Grade
Toxic
Corrosive
Toluene-α-sulphonyl fluoride ≥99.0%, Proteomics Grade
Catalog # M145-25G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Corrosive
Toluene-α-sulphonyl fluoride ≥99.0%, Proteomics Grade
Catalog # M145-25G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Identification:
    PASS
  • Melting Point:
    91 - 95 °C
  • Purity:
    99.0 %
  • Solubility (5 %, EtOH):
    PASS
  • TLC:
    ONE SPOT

Specification Test Results

About this item

 :
 :174.2 g/mol
Boiling Pt:80…85 °C (0.3 Torr)
Melting Pt:91…92 °C
Storage Temperature:Ambient
EINECS:206-350-2
UN:2923
ADR:8 (6.1),III
 :GHS06
 :GHS05
CAS Number:329-98-6
Formula:C₇H₇FO₂S
MDL Number:MFCD00007424