Order Entry
Japan
ContactUsLinkComponent
Bismuth(III) carbonate basic Ph. Eur.
Bismuth(III) carbonate basic Ph. Eur.
Catalog # 83518.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bismuth(III) carbonate basic Ph. Eur.
Catalog # 83518.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay (Bi) (calculated on dried):
    80.0 - 82.5 %
  • Appearance:
    White/almost white powder
  • Identification (CO₃):
    Passes test
  • Identification (Bi):
    Passes test
  • Appearance of solution:
    Passes test
  • Cl (Chloride):
    500 ppm
  • NO₃ (Nitrate):
    0.4 %
  • Alkali and alkaline-earth metals:
    1.0 %
  • Loss on drying (105°C):
    1.0 %
  • Residual solvents:
    Unlikely by manuf.process
  • Residual metal catalysts (skip lot)::
    .
  • As (Arsenic):
    5 ppm
  • Cd (Cadmium):
    0.15 ppm
  • Co (Cobalt):
    1.5 ppm
  • Hg (Mercury):
    2.4 ppm
  • Ni (Nickel):
    6 ppm
  • Pb (Lead):
    20 ppm
  • V (Vanadium):
    10 ppm
  • Conforms to Ph.Eur.:
    Passes test

Specification Test Results

About this item

 :
 :509.97 g/mol
Merck Index:13,01283
Density:6.86 g/cm³ (25 °C)
EINECS:227-567-9
CAS Number:5892-10-4
Formula:(BiO)₂CO₃
MDL Number:MFCD00010877