L(+)-Serine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Catalog # 2088-06
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Meets B.P. Chemical Specifications:98.5 - 101.5%
- Meets U.S.P Requirements:Passes Test
- GMP Manufactured Product:14.0 - 15.6 Degree
- Meets E.P. Chemical Specifications:≤0.2%
- Store in well-closed,light-resistant containers:≤0.1%
- Preserve in well-closed containers.:≤0.05%
- CAUTION: For Manufacturing, processing or repackaging:≤0.03%
- Bulk Pharmaceutical Chemical:≤0.003%
- USP - Assay (C₃H₇NO₃) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):14.0 - 15.6 Degree
- USP - Loss on Drying at 105°C:≤0.2 %
- USP - Residue on Ignition:≤0.1 %
- USP - Chloride (Cl):≤0.05 %
- USP - Sulfate (SO₄):≤0.03 %
- USP - Iron (Fe):≤0.003 %
- USP - Related Compounds - Individual Impurities:≤0.5 %
- USP - Related Compounds - Total Impurities:≤2.0 %
- EP/BP - Assay (C₃H₇NO₃) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):14.0 - 16.0 Degree
- EP/BP-Ninhydrin-Positive Substances-Each:≤0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤0.5 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Sulfate (SO₄):≤300 ppm
- EP/BP - Ammonium (NH₄):≤0.02 %
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
Specification Test Results
About this item
:
:105.09 g/mol
Merck Index:14,00846
Boiling Pt:259 °C (1013 hPa)
Melting Pt:215…225 °C
Density:1.6 g/cm³ (22 °C)
Storage Temperature:Ambient
EINECS:200-274-3
CAS Number:56-45-1
Formula:C₃H₇NO₃
MDL Number:MFCD00064224