ARISTAR®VWR® , Indium Standard Solution, 10000 mg/L In in dilute Nitric Acid (from In), Standard for ICP
Catalog # 455492J
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Indium
- Symbol:In
Specifications
About this item
In in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.