Order Entry
Japan
ContactUsLinkComponent
VWR® ARISTAR®, Chloride Standard Solution, 200 mg/L Cl- in Water (from NaCl) Standard for Ion Chromatography
VWR® ARISTAR®, Chloride Standard Solution, 200 mg/L Cl- in Water (from NaCl) Standard for Ion Chromatography
Catalog # 458082H
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Chloride Standard Solution, 200 mg/L Cl- in Water (from NaCl) Standard for Ion Chromatography
Catalog # 458082H
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    200 mg/l
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Chloride standard solution
  • Contents:
    NaCl in H₂O

Specifications

About this item

(Cl in H₂O).

Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory

Supplied with certificate of analysis.