di-Sodium β-glycerophosphate pentahydrate ≥98%, GPR RECTAPUR®
27874.295
:
- パックタイプ:Plastic bottle
- Pk:1 kg
- Seq:4024
- Assay (calculated on anhydrous):Min. 98 %
- Alkalinity:Max. 0.15 meq/g
- Heavy metals (as Pb):Max. 20 ppm
- Water:25 - 35 %
- PO4 (Phosphate):Max. 0.1 %
:
:306.11 g/mol
:212-464-3
:13408-09-8
:819-83-0
:C₃H₇Na₂O₆P·5H₂O
:MFCD00002135