VWR® Penicillinase (beta-Lactamase)
390843G
:
- Pack type:Vial
- Pk:1 Vial
- CAS No.:9001-74-5
- Storage conditions:Refrigerator
- Enzyme name:Penicillinase (beta-Lactamase)
A freeze-dried product with >3000 ß-l units per vial.
One unit of ß-l activity is defined as the amount of enzyme that will catalyse the hydrolysis of 1,0 µm of benzylpenicillin per minute at 25 °C and pH 7,0.