VWR®, α-Endosulfan Certified Reference Material
α-Endosulfan Certified Reference Material, Glass ampoule, 10 mg
- Pack type:Glass ampoule
- Pk:10 mg
- Description:α-Endosulfan Certified Reference Material
- Cat. No.:124082E
- Formula:C₉H₆Cl₆O₃S
- Description:α-Endosulfan
- Storage conditions:Ambient (>5 °C)
- Molecular weight:406.93 g/mol
- CAS No.:959-98-8



