Order Entry
Japan
ContactUsLinkComponent
VWR® Phase Separating Paper, Grade 511
VWR® Phase Separating Paper, Grade 511
Supplier:  VWR Collection
Certificates
undefined
VWR® Phase Separating Paper, Grade 511
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 516-0311
  • 516-0313
  • 516-0315
  • 516-0314
  • 516-0312
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • Filtration speed:
  • 200 s (Herzberg)
  • 200 s (Herzberg)
  • 200 s (Herzberg)
  • 200 s (Herzberg)
  • 200 s (Herzberg)