Order Entry
Japan
ContactUsLinkComponent
 
VWR®, Bead Mill Homogeniser
  432-0367
undefined
VWR®, Bead Mill Homogeniser
  432-0367

 

  • Capacity:
    Max. 16 tubes: 12×2 ml tubes + 4×7 ml tubes
  • Description:
    Bead Mill homogeniser, 230 V
  • Seq:
    001
  • Model:
    Bead Mill homogeniser
  • Application areas:
    Sample preparation, DNA, RNA, Protein extraction
  • Speed range:
    0,8 to 6 m/s
  • Sample volume:
    25 µl - 5 ml
  • Run time:
    1 s - 10 min
  • No. of cycles:
    1 - 10
  • Weight:
    24 kg
  • W×D×H:
    305×432×368 mm
  • Pk:
    1

 

 

The VWR® Bead Mill homogeniser is a part of the newest generation of Bead Mill homogenisers. This economical system is specifically designed for grinding, lysing and homogenisation of up to 16 samples. Using sample tubes pre-filled with a variety of lysing beads, the VWR Bead Mill vigorously and uniformly shakes the tubes providing an efficient, consistent high yield and quality homogenisation usually in less than 40 seconds.

  • Capacity for 16 samples (12×2 ml tubes plus 4×7 ml tubes, 16 in total)
  • Run time: 1 second to 10 minutes
  • 0,8 to 6 m/s
  • Up to 99 programs

The VWR® Bead Mill simultaneous homogenises 12×0,5, 4×1,5, 12×2 or 4×7 ml samples.

The VWR® Bead Mill is an ideal solution for releasing DNA, RNA proteins and enzymes from tough samples while still retaining molecular integrity.

It does not require a cool down period between runs, allowing non stop use and high throughput. The unit features a convenient front loading design with snap-in tube carriage.

 : Drive unit is supplied with tube carriage, user manual and 2 year warranty. VWR® disposable bead tubes must be ordered separately.

Grinding mix