Order Entry
Japan
ContactUsLinkComponent
Amyl acetate ≥98.5% (by GC), GPR RECTAPUR®
Flammable
Amyl acetate ≥98.5% (by GC), GPR RECTAPUR®
Catalog # 85737.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Amyl acetate ≥98.5% (by GC), GPR RECTAPUR®
Catalog # 85737.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Assay (GC):
    min. 98.5 %
  • Density (d20°C/ 4°C):
    0.875 - 0.877
  • Identity (IR):
    Passes test

Specification Test Results

About this item

 :
 :130.19 g/mol
Storage Temperature:Ambient
UN:1104
ADR:3,III
 :GHS02
CAS Number:628-63-7
Formula:C₇H₁₄O₂
MDL Number:MFCD00009500