Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Japan
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Citric acid monohydrate
Citric acid monohydrate, Rankem®
Citric acid monohydrate, Rankem®
Citric acid monohydrate, Rankem®
Catalog # RANK0770-06
Supplier:
Transene
CAS Number:
Expand image
Citric acid monohydrate, Rankem®
Catalog # RANK0770-06
Supplier:
Transene
CAS Number:
Specifications
Pk:
25 kg
Seq:
4283
Specifications
About this item
Product Details
:
:
210.14 g/mol
CAS Number:
5949-29-1
Formula:
C₆H₈O₇.H₂O
MDL Number:
MFCD00149972
Product Details