CyMet BS3 VET for hematology CN-free, J.T.Baker®
Catalog # 2975.0500PE
Supplier: Avantor
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Performance Test *:passes test
- Conductivity (mS):12.80 - 13.30
- Osmolality (mOsm/kg):312 - 322
- pH at 25°C:6.30 - 6.90
Specification Test Results
About this item
:
Storage Temperature:Refrigerator
:1