Diluid III Diff for hematology, J.T.Baker®
Catalog # 3963.
Supplier: Avantor
CAS Number:
Specifications
- Pk:20 L
- Seq:5302
Specifications
Specification Test Results
- Conductivity (mS):16.00 - 21.00
- Osmolality (mOsm/kg):325 - 335
- Particles 3 - 30 µ:max. 2
- Performance Test:passes test
- pH at 25°C:6.90 - 7.10
- Total Bacterial Growth (CFU/100ml):max. 5
Specification Test Results
About this item
:
Storage Temperature:Refrigerator
:1