Guanidinium hydrochloride low ash, Technical Grade without anti-caking agent
Catalog # VWRAE424-5KG
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:5 kg
- Seq:4114
Specifications
Specification Test Results
- Ammonium chloride:Max. 0.5%
- Ash:Max. 0.4%
- Moisture (KF):Max. 0.3%
- Purity:Min. 98.5%
Specification Test Results
About this item
:
:95.53 g/mol
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026