
N,N-Diethyl-p-phenylenediammonium sulphate ≥99%, AnalaR NORMAPUR® analytical reagent
Catalog # 87026.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 0,05 %
- Water:Max. 0,5 %
- Fe (Iron):Max. 10 ppm
Specification Test Results
About this item
:
:262.33 g/mol
Boiling Pt:274…275 °C (1013 hPa)
Melting Pt:184…186 °C
EINECS:228-500-6
:GHS07
Storage Temperature:Ambient
CAS Number:6283-63-2
Formula:C₁₀H₁₈N₂O₄S
MDL Number:MFCD00012993