Rankem®, Specific Gravity Bottles, Class A
: Transene
Specific Gravity Bottle Class A with Ground in Thermometer range 0 to 50 °C with work Certificate (Pyknometer). Suitable for determining the density with built in thermometer.
Frequently Bought Together