Order Entry
Japan
ContactUsLinkComponent
Water, sterile filtered
Water, sterile filtered
Catalog # AX021403
Supplier:  VWR Chemicals
CAS Number:  
undefined
Water, sterile filtered
Catalog # AX021403
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    12
  • Seq:
    0012
  • Pk Info:
    12x290 ml

Specifications

About this item

 :
 :18.02 g/mol
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332