Order Entry
Japan
ContactUsLinkComponent
Disodium hydrogen phosphate, anhydrous 98.0-100.5%, Multi-Compendial, Macron Fine Chemicals™
Disodium hydrogen phosphate, anhydrous 98.0-100.5%, Multi-Compendial, Macron Fine Chemicals™
Catalog # MACR7810-06
CAS Number:  
Disodium hydrogen phosphate, anhydrous 98.0-100.5%, Multi-Compendial, Macron Fine Chemicals™
Catalog # MACR7810-06
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • USP - (Na2HPO4) (dried basis):
    98.00 to 100.50 %
  • USP - Identification :
    Passes Test
  • USP - Loss on Drying at 130°C:
    Max.5.0 %
  • USP - Insoluble Substances:
    Max.0.4 %
  • USP - Chloride (Cl):
    Max.0.06 %
  • USP - Sulphate (SO₄):
    Max.0.2 %
  • USP - Arsenic (As):
    Max.16 ppm
  • USP - Heavy Metals (as Pb):
    Max. 20 ppm
  • BP/EP - Identification A :
    Passes Test
  • BP/EP - Identification B :
    Passes Test
  • BP/EP - Identification C :
    Passes Test
  • BP/EP - Identification D :
    Passes Test
  • BP/EP - Appearance of Solution :
    Passes Test
  • BP/EP - Arsenic (As):
    Max. 2 ppm
  • BP/EP - Heavy Metals (as Pb):
    Max. 10 ppm
  • BP/EP - Iron (Fe):
    Max.20 ppm
  • BP/EP - Chloride (Cl):
    Max.500 ppm
  • BP/EP - Sulphate (SO₄):
    Max.500 ppm
  • BP/EP - Reducing Substances :
    Passes Test
  • BP/EP - Monosodium Phosphate:
    Max.2.5 %
  • BP/EP - Loss on Drying at 105°C:
    Max.1.0 %
  • BP/EP - Assay (Na2HPO4) (dried basi :
    98.00 - 101.00 %

Specification Test Results

About this item

 :
 :141.96 g/mol
CAS Number:7558-79-4
Formula:Na₂HPO₄
MDL Number:MFCD00003496