Order Entry
Japan
ContactUsLinkComponent
VWR®, External PT100 Probe for Chillers
  462-0553
VWR®, External PT100 Probe for Chillers
  462-0553
  • Description:
    External PT 100 probe for chillers, with 8 m cable length
  • Pk:
    1
External PT100 probe is used for with chillers and coolers, 6000 series chiller, non-refrigerated coolers, 4100 liquid-to-liquid coolers.