Rankem®, Burettes, PTFE Key Stopcock, Rotaflow, Class B
Supplier: Transene
Certificates
About this item
Burettes fitted with Rotaflow general purpose screw thread stopcock with PTFE key.
- Made of borosilicate glass
- Graduated
Certifications: Accuracy as per Class B of IS 1997:2008, ISO 385:2005.
Frequently Bought Together