Order Entry
Japan
Orders LinkContactUsLinkComponent
J.T.Baker®, ProClean cleaners
J.T.Baker®, ProClean cleaners
ProClean cleaner, 5 L
Catalog # 3900.
Supplier:   Avantor
J.T.Baker®, ProClean cleaners
Catalog # 3900.
Supplier:  Avantor
CAS Number:  
J.T.Baker®, ProClean cleaners
ProClean cleaner, 5 L
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 5 L
Product Details & Documents
Caution: pH is measured directly after production. In course of time the actual value will decrease. This will not influence the performance of the reagent.
Specifications
Specifications
  • Pk:
    5 L
  • Description:
    ProClean cleaner