Order Entry
Japan
ContactUsLinkComponent
LiChrospher® RP-18E, HPLC Columns
LiChrospher® RP-18E, HPLC Columns
Supplier:  HICHROM
   Sustainable Options Available
Certificates
undefined
LiChrospher® RP-18E, HPLC Columns
Supplier:  HICHROM
   Sustainable Options Available
Specifications
  • Catalog No:
  • SPRP18E510C5SK
  • LISPRP18E5150M
  • LISPRP18E10250
  • LISPRP18E5200A
  • LISPRP18E-5-250A
  • LISPRP18E5250F
  • LISPRP18E5150S
  • LISPRP18E-5-125A
  • LISPRP18E-5-10C5
  • LISPRP18E-5-150A
  • LISPRP18E-5-50AF
  • LISPRP18E5125F
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 21,6%
  • 21,6%
  • 21,6%
  • 21,6%
  • 21,6%
  • 21,6%
  • 21,6%
  • 21,6%
  • 21,6%
  • 21,6%
  • 21,6%
  • 21,6%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • 350 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)