VWR®, Bottles, Rectangular, Amber, HDPE, Wide Mouth
Supplier: VWR Collection
Certificates
About this item
Space-saving, leakproof amber bottles are for sampling, storage, or shipment of light-sensitive dry chemicals and liquids.
- HDPE construction
- Amber tint protects against UV radiation
- Ideal for holding light-sensitive dry chemicals and liquids
Bottles comply with current USP requirements for maximum light transmission. Sturdy construction prevents bulging, translucent material allows for optical clarity, and wide mouth makes filling easy. Bottles have excellent chemical resistance to most acids, bases, and alcohols, and can be stored in freezer to –100 °C.
Bottle VWRU414004-200 has integrated handle for better grip, greater safety, and pouring convenience.
Delivery: Amber polypropylene screw caps included.
Frequently Bought Together