Rankem®, Serological Pipettes, Graduated, Class B
Supplier: Transene
Certificates
About this item
Borosilicate glass pipettes, class B.
- Colour coded
- Graduated upto tip
- Calibrated to deliver their total capacity
Delivery: Supplied with certificate.
Frequently Bought Together