Rankem®, Condensers, Graham
Supplier: Transene
Certificates
About this item
These Graham condensers are coiled distillate type and have drip tip, interchangeable inner and outer joints.
Frequently Bought Together
These Graham condensers are coiled distillate type and have drip tip, interchangeable inner and outer joints.