Rankem®, Measuring Pipettes (Mohr), Class A
: Transene
Borosilicate glass pipettes, class A.
- Colour coded
- Graduated for delivery from zero mark upto the graduation mark
: Supplied with certificate.
Frequently Bought Together
Borosilicate glass pipettes, class A.