Rankem®, Flasks, Round Bottom, Glass, Clear, Standard Gound Joints, Two Necks
Catalog # RANKFBRB2N-0500-AS
Supplier: Transene
Specifications
- Capacity:500 ml
- Seq:03
- Pk:4
- Side necks:19/26 NS
- Centre neck:24/29 NS
Specifications
About this item
Borosilicate glass, clear, with one centre neck and one angled side neck.
- Side neck with an interchangeable joint