EDTA disodium salt dihydrate, GenAR® USP, Macron Fine Chemicals™
Catalog # 7565-21
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:55 lb
- Seq:7140
Specifications
Specification Test Results
- USP - Assay (Dried Basis):99.0 - 101.0 %
- Appearance (white crystals or white powder):Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - pH (1 in 20):4.0 - 6.0
- USP - Loss on Drying:8.7 - 11.4 %
- USP - Calcium (Ca):Passes Test
- USP - Limit of Nitrilotriacetic Acid:≤0.1 %
- EP - Assay:98.5 - 101.0 %
- EP - Appearance of Solution:Passes Test
- EP - Identification A:Passes Test
- EP - Identification B:Passes Test
- EP - Identification D:Passes Test
- EP - pH....:4.0 - 5.5
- EP - Iron (Fe):≤80 ppm
- EP - Impurity A:≤0.1 %
- JP - Assay:99.0 - 101.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Identification C:Passes Test
- JP - pH:4.3 - 4.7
- JP - Clarity and Color of Solution:Passes Test
- JP - Cyanide (CN):Passes Test
- JP - Heavy Metals:≤10 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Residue on Ignition:37.0 - 39.0 %
Specification Test Results
About this item
:
:372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037