Nessler reagent, StandARd® for ammonia nitrogen determination, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7783-33-7
MDL Number:MFCD00049656
Merck Index:12,07862
Boiling Pt:356.6 °C (1013 hPa)
Melting Pt:–38.9 °C
Density:13.6 g/cm³ (20 °C)
EINECS:231-990-4
UN:3289
ADR:6.1 (8),II
:GHS05
:GHS06
:GHS08
:GHS09
Specification Test Results
- Suitability:Passes Test
Specification Test Results
Frequently Bought Together