Sodium thiosulphate 0.01 N (N/100), StandARd® volumetric solution, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7772-98-7
:
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
:158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
Specification Test Results
- Normality:0.0095 - 0.0105
Specification Test Results
Frequently Bought Together