


Ammonia solution 28 - 30% (as NH₃) NF, ACS, Macron Fine Chemicals™
Catalog # 3261-56
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:1,998 L
- Seq:5861
Specifications
Specification Test Results
- NF - Assay (NH₃)(NF):27.0 - 31.0 %
- NF - Identification:Passes Test
- NF - Nonvolatile Residue:≤0.05 %
- NF - Readily Oxidizable Substances:Passes Test
- ACS - Assay (as NH₃):28.0 - 30.0 %
- Appearance:Passes Test
- ACS - Carbon Dioxide (CO₂):≤0.002 %
- ACS - Residue after Ignition:≤0.002 %
- ACS - Substances Reducing Permanganate:Passes Test
- Arsenic and Antimony (as As):≤3 ppm
- Chloride (Cl):≤0.5 ppm
- Trace Impurities - Heavy Metals:≤0.5 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- Sulfate (SO₄):≤2 ppm
- Trace Impurities - Aluminum (Al):≤0.300 ppm
- Trace Impurities - Boron (B):≤0.100 ppm
- Trace Impurities - Calcium (Ca):≤0.300 ppm
- Trace Impurities - Chromium (Cr):≤0.200 ppm
- Trace Impurities - Copper (Cu):≤0.10 ppm
- Trace Impurities - Gold (Au):≤0.300 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Trace Impurities - Lead (Pb):≤0.200 ppm
- Trace Impurities - Magnesium (Mg):≤0.300 ppm
- Trace Impurities - Manganese (Mn):≤0.200 ppm
- Trace Impurities - Nickel (Ni):≤0.1 ppm
- Trace Impurities - Potassium (K):≤0.300 ppm
- Trace Impurities - Sodium (Na):≤2.000 ppm
- Trace Impurities - Tin (Sn):≤0.300 ppm
- Trace Impurities - Titanium (Ti):≤0.300 ppm
- Trace Impurities - Zinc (Zn):≤0.300 ppm
Specification Test Results
About this item
:
:35.05 g/mol
UN:2672
ADR:8,III
:GHS05
:GHS07
:GHS09
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650