


Phenolphthalein alcoholic solution, BAKER ANALYZED® USP Test Solution (TS), J.T.Baker®
Catalog # 5927-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Visual Transition Interval - pH…8.0 (Colorless):Passes Test
- Visual Transition Interval - pH…10.0 (Pink):Passes Test
Specification Test Results
About this item
:
:318.33 g/mol
Flash Pt:13 °C
UN:1170
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913