Order Entry
Japan
ContactUsLinkComponent
Cyclohexane ≥99.0% (by GC), AR® ACS, Macron Fine Chemicals™
Flammable
Health
Irritant
Environment
Cyclohexane ≥99.0% (by GC), AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Flammable
Health
Irritant
Environment
Cyclohexane ≥99.0% (by GC), AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥99.0%
  • ACS - Assay (C₆H₁₂) (by GC, corrected for water):
    ≥99.0 %
  • Appearance:
    Passes Test
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤0.001 %
  • ACS - Substances Darkened by H₂SO₄:
    Passes Test
  • ACS - Water (H₂O)(by KF, coulometric):
    ≤0.02 %

Specification Test Results

Related Information

Frequently Bought Together