Hydrochloric acid 0.1 N (N/10), StandARd® volumetric solution, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
:36.46 g/mol
:GHS05
:GHS07
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:0.0995 - 0.1005
- Normality:0.0995 - 0.1005
Specification Test Results
Related Information
Frequently Bought Together