Acetone ≥99.5%, AR® ACS, Macron Fine Chemicals™
Catalog # MACR2443-10
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth, PVC-coated
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.5%
- For Laboratory,Research,or Manufacturing Use:≤0.002%
- ACS - Assay (CH₃COCH₃) (by GC, corrected for water):≥99.5 %
- ACS - Aldehydes (as HCHO):≤0.002 %
- ACS - Color (APHA):≤10
- ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):≤0.05 %
- ACS - Methanol (CH₃OH)(by GC):≤0.05 %
- ACS - Residue after Evaporation:≤0.001 %
- ACS - Solubility in H₂O:Passes Test
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Titrable Acid (meq/g):≤0.0003
- ACS - Titrable Base (meq/g):≤0.0006
- ACS - Water (H₂O):≤0.5 %
Specification Test Results
About this item
:
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:≤20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS02
:GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765