Silica gel, BAKER ANALYZED® 80-200 Mesh, J.T.Baker®
Catalog # 3404-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, wide mouth
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≤ 13 %
- Loss on Ignition:≤13 %
- Alcohol-Ether Soluble Substances:≤0.05 %
- Flow Rate, mL/min.:≥0.5
- Mesh - Thru U.S. No. 80 Sieve:≥90.0 %
- Mesh - Thru U.S. No. 200 Sieve:≤10.0 %
Specification Test Results
About this item
:
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1710 °C
Density:2.10 kg/m³
Storage Temperature:Ambient
REACH:01-2119379499-16
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232